siRNA Details
| Gene Symbol: Tmprss9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081688.2 | NP_001075157.2 | 12 | 1913 | ||
| GenBank mRNA | BC158102.1 | - | 12 | 2009 | 9007387 | |
| BC172169.1 | - | 12 | 2009 | 9054839 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss9| Gene Symbol: Tmprss9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081688.2 | NP_001075157.2 | 12 | 1913 | ||
| GenBank mRNA | BC158102.1 | - | 12 | 2009 | 9007387 | |
| BC172169.1 | - | 12 | 2009 | 9054839 | ||