siRNA Details
| Gene Symbol: Tmprss9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081688.2 | NP_001075157.2 | 1 | 18 | ||
| GenBank mRNA | AB109391.1 | - | 1 | 129 | ||
| BC158102.1 | - | 1 | 114 | 9007387 | ||
| BC172169.1 | - | 1 | 114 | 9054839 | ||

Tmprss9
Genomic Map