siRNA Details
| Gene Symbol: E130309D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013784.1 | NP_001013806.1 | 3 | 428 | ||
| GenBank mRNA | AK143211.1 | - | 3 | 429 | ||
| BC150886.1 | - | 3 | 323 | 9087797 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130309D14Rik| Gene Symbol: E130309D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013784.1 | NP_001013806.1 | 3 | 428 | ||
| GenBank mRNA | AK143211.1 | - | 3 | 429 | ||
| BC150886.1 | - | 3 | 323 | 9087797 | ||