siRNA Details
| Gene Symbol: Trim80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033775.3 | NP_001028947.1 | 3 | 189 | ||
| GenBank mRNA | AK133415.1 | - | 3 | 190 | ||
| BC147608.1 | - | 3 | 207 | 9056598 | ||
| BC157912.1 | - | 3 | 207 | 9088129 | ||

Trim80
Genomic Map