siRNA Details
| Gene Symbol: Spink11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048217.3 | NP_001041682.2 | 4 | 212 | ||
| GenBank mRNA | AK136658.1 | - | 1 | 655 | ||
| AK136741.1 | - | 4 | 210 | |||
| BC147524.1 | - | 4 | 181 | 9056516 | ||
| BC147541.1 | - | 4 | 181 | 9056532 | ||
| DQ437331.1 | - | 4 | 212 | |||

Spink11
Genomic Map