siRNA Details
| Gene Symbol: Sgpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004173.2 | NP_001004173.1 | 2 | 302 | ||
| GenBank mRNA | BC049995.1 | - | 2 | 303 | 6399201 | |
| BC147843.1 | - | 2 | 491 | 9087560 | ||
| BC147850.1 | - | 2 | 491 | 9087567 | ||
| BY234834.1 | - | 2 | 283 | |||
| BY297240.1 | - | 2 | 301 | |||

Sgpp2
Genomic Map