siRNA Details
| Gene Symbol: Creg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011804.2 | NP_035934.1 | 1 | 223 | ||
| GenBank mRNA | AF084524.1 | - | 1 | 223 | ||
| AK005388.1 | - | 1 | 244 | |||
| AK010947.1 | - | 1 | 240 | |||
| AK085844.1 | - | 1 | 35 | |||
| AK144101.1 | - | 1 | 242 | |||
| AK150006.1 | - | 1 | 195 | |||
| AK181270.1 | - | 1 | 28 | |||
| BC027426.1 | - | 1 | 223 | 3670649 | ||

Creg1
Genomic Map