siRNA Details
| Gene Symbol: Gm1965 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136090.1 | - | 3 | 542 | ||
| AK136244.1 | - | 2 | 439 | |||
| AK139519.1 | - | 3 | 555 | |||
| AK165099.1 | - | 3 | 539 | |||
| BC147526.1 | - | 3 | 459 | 9056518 | ||
| BC147534.1 | - | 3 | 458 | 9056525 | ||
| BC172072.1 | - | 3 | 459 | 9054358 | ||

Gm1965
Genomic Map