siRNA Details
Gene Symbol: Ccdc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101535.1 | NP_001095005.1 | 1 | 278 | ||
GenBank mRNA | AK133673.1 | - | 311 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101535.1 | NP_001095005.1 | 1 | 278 | ||
GenBank mRNA | AK133673.1 | - | 311 |