siRNA Details
| Gene Symbol: Gm1966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277179.1 | NP_001264108.1 | 3 | 2146 | ||
| NM_001277180.1 | NP_001264109.1 | 3 | 2158 | |||
| NM_001277181.1 | NP_001264110.1 | 4 | 2230 | |||
| NM_001277182.1 | NP_001264111.1 | 4 | 2296 | |||
| NM_001277183.1 | NP_001264112.1 | 6 | 2430 | |||
| GenBank mRNA | AK156879.1 | - | 4 | 2212 | ||
| AK171325.1 | - | 3 | 2160 | |||

Gm1966
Genomic Map