siRNA Details
| Gene Symbol: Rhox8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004193.2 | NP_001004193.1 | 2 | 228 | ||
| GenBank mRNA | AY329370.1 | - | 2 | 228 | ||
| BC139098.1 | - | 2 | 211 | 8862120 | ||
| DQ058645.1 | - | 3 | 237 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhox8| Gene Symbol: Rhox8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004193.2 | NP_001004193.1 | 2 | 228 | ||
| GenBank mRNA | AY329370.1 | - | 2 | 228 | ||
| BC139098.1 | - | 2 | 211 | 8862120 | ||
| DQ058645.1 | - | 3 | 237 | |||