siRNA Details
| Gene Symbol: Iigp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013828.2 | NP_001013850.1 | 688 | |||
| GenBank mRNA | BC020118.1 | - | 689 | 3675329 | ||
| BC029756.1 | - | 692 | 4013233 | |||
| BC040796.1 | - | 702 | 3495064 | |||

Iigp1b
Genomic Map