siRNA Details
| Gene Symbol: Ldlrad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033979.1 | NP_001029151.1 | 3 | 372 | ||
| GenBank mRNA | AK132881.1 | - | 3 | 372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ldlrad2| Gene Symbol: Ldlrad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033979.1 | NP_001029151.1 | 3 | 372 | ||
| GenBank mRNA | AK132881.1 | - | 3 | 372 | ||