siRNA Details
| Gene Symbol: Fbxl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176959.3 | NP_795933.2 | 2 | 600 | ||
| GenBank mRNA | AK048688.1 | - | 2 | 614 | ||
| AK051849.1 | - | 2 | 619 | |||
| AK084174.1 | - | 1 | 48 | |||
| AK129227.1 | - | 2 | 548 | |||
| BC050864.1 | - | 2 | 554 | |||
| BC091646.1 | - | 2 | 600 | 30355382 | ||

Fbxl7
Genomic Map