siRNA Details
| Gene Symbol: Taar9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010831.1 | NP_001010831.1 | 1 | 168 | ||
| GenBank mRNA | BC139084.1 | - | 1 | 217 | 8862106 | |
| BC139085.1 | - | 1 | 216 | 8862107 | ||
More Information
Set Membership: |
|

Taar9
Genomic Map