siRNA Details
| Gene Symbol: Chrna10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081424.1 | NP_001074893.1 | 2 | 549 | ||
| GenBank mRNA | AK033068.1 | - | 2 | 549 | ||
| BC147245.1 | - | 2 | 264 | 9056293 | ||
| BC147246.1 | - | 2 | 264 | 9056310 | ||
| BC151033.1 | - | 2 | 264 | 9087946 | ||
| BC151035.1 | - | 2 | 264 | 9087948 | ||

Chrna10
Genomic Map