siRNA Details
| Gene Symbol: Nox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285833.1 | NP_001272762.1 | 14 | 1411 | ||
| NM_001285835.1 | NP_001272764.1 | 12 | 1294 | |||
| NM_015760.5 | NP_056575.1 | 12 | 1298 | |||
| GenBank mRNA | AB041034.1 | - | 12 | 1149 | ||
| AB042745.1 | - | 12 | 1149 | |||
| AF218723.1 | - | 12 | 1149 | |||
| AF261944.1 | - | 12 | 1153 | |||
| AF276957.1 | - | 12 | 1160 | |||
| AK050371.1 | - | 12 | 1153 | |||
| AK052905.1 | - | 12 | 1149 | |||
| AK085509.1 | - | 12 | 1154 | |||
| AK149398.1 | - | 12 | 1149 | |||
| AK149455.1 | - | 12 | 1149 | |||
| AK149492.1 | - | 12 | 1149 | |||
| BC021378.1 | - | 14 | 1259 | |||
| BC138918.1 | - | 12 | 1145 | 8861940 | ||
| BC145462.1 | - | 12 | 1141 | |||
| EU861989.1 | - | 3 | 180 | |||

Nox4
Genomic Map