siRNA Details
| Gene Symbol: E2f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033270.2 | NP_150373.2 | 4 | 713 | ||
| GenBank mRNA | AB221552.1 | - | 4 | 474 | ||
| AF032131.1 | - | 4 | 691 | |||
| AK035116.1 | - | 4 | 710 | |||
| AK146792.1 | - | 5 | 775 | |||
| AK149061.1 | - | 869 | ||||
| AK183289.1 | - | 1 | 63 | |||
| AK187122.1 | - | 1 | 58 | |||
| BC019166.1 | - | 5 | 775 | 4236246 | ||
| BC037656.1 | - | 5 | 656 | 5358554 | ||
| BC057929.1 | - | 4 | 701 | 5289475 | ||

E2f6
Genomic Map