siRNA Details
| Gene Symbol: Ebi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015766.2 | NP_056581.1 | 3 | 539 | ||
| GenBank mRNA | AF013114.1 | - | 3 | 522 | ||
| AK155023.1 | - | 3 | 502 | |||
| AK155903.1 | - | 3 | 394 | |||
| BC008209.1 | - | 3 | 487 | 3154957 | ||
| CX240802.1 | - | 3 | 486 | |||

Ebi3
Genomic Map