siRNA Details
| Gene Symbol: Sacs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172809.3 | NP_766397.2 | 8 | 2355 | ||
| GenBank mRNA | AK038655.1 | - | 2 | 1919 | ||
| AK046501.1 | - | 7 | 2210 | |||
| AK053626.1 | - | 7 | 1790 | |||
| AK160797.1 | - | 8 | 2356 | |||
| BC138482.1 | - | 7 | 1911 | 8861502 | ||
| BC138483.1 | - | 7 | 1911 | 8861503 | ||
| BC171956.1 | - | 7 | 1911 | 9054242 | ||

Sacs
Genomic Map