siRNA Details
| Gene Symbol: Sap30l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081168.1 | NP_001074637.1 | 3, 4 | 867 | ||
| GenBank mRNA | AK010234.1 | - | 3, 4 | 355 | ||
| AK203234.1 | - | 1, 2 | 55 | |||
| BC051686.1 | - | 3, 4 | 867 | 6433799 | ||
| BC094930.1 | - | 4, 5 | 586 | |||
| BC117029.1 | - | 3, 4 | 581 | 40126348 | ||
| BC117031.1 | - | 3, 4 | 581 | 40126350 | ||

Sap30l
Genomic Map