siRNA Details
| Gene Symbol: Fbxl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015794.1 | NP_056609.1 | 2, 3 | 1687 | ||
| GenBank mRNA | AF176529.1 | - | 2, 3 | 407 | ||
| AK154497.1 | - | 2, 3 | 196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxl17| Gene Symbol: Fbxl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015794.1 | NP_056609.1 | 2, 3 | 1687 | ||
| GenBank mRNA | AF176529.1 | - | 2, 3 | 407 | ||
| AK154497.1 | - | 2, 3 | 196 | |||