siRNA Details
| Gene Symbol: Fbxo17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015796.2 | NP_056611.2 | 4 | 917 | ||
| GenBank mRNA | AF176532.1 | - | 4 | 834 | ||
| AK133408.1 | - | 4 | 877 | |||
| AK134284.1 | - | 3 | 1411 | |||
| BC026470.1 | - | 4 | 903 | 4486850 | ||

Fbxo17
Genomic Map