siRNA Details
Gene Symbol: Crim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015800.3 | NP_056615.1 | 5, 6 | 1522 | ||
GenBank mRNA | AF168680.1 | - | 5, 6 | 974 | ||
AK053197.1 | - | 3, 4 | 465 | |||
BC100348.1 | - | 5, 6 | 1307 | 30521148 |
