siRNA Details
| Gene Symbol: Slc25a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177572.1 | NP_001171043.1 | 8 | 983 | ||
| NM_015829.3 | NP_056644.1 | 8 | 983 | |||
| GenBank mRNA | AF164632.1 | - | 8 | 928 | ||
| AK002829.1 | - | 8 | 858 | |||
| AK012670.1 | - | 8 | 874 | |||
| AK029020.1 | - | 8 | 914 | |||
| AK150496.1 | - | 8 | 929 | |||
| AK151180.1 | - | 8 | 929 | |||
| AK152237.1 | - | 8 | 929 | |||
| BC016571.1 | - | 2 | 120 | 3600909 | ||

Slc25a13
Genomic Map