siRNA Details
| Gene Symbol: Il17rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019583.3 | NP_062529.2 | 11 | 1255 | ||
| GenBank mRNA | AF208108.1 | - | 11 | 1168 | ||
| AF208109.1 | - | 11 | 1794 | |||
| AK045627.1 | - | 11 | 1256 | |||
| AK050139.1 | - | 11 | 1176 | |||
| BC026546.1 | - | 11 | 1155 | 5042466 | ||

Il17rb
Genomic Map