siRNA Details
| Gene Symbol: Galns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016722.4 | NP_057931.3 | 11 | 1314 | ||
| GenBank mRNA | AA793622.1 | - | 3 | 234 | ||
| AF111346.1 | - | 11 | 1235 | |||
| AK159592.1 | - | 11 | 1291 | |||
| AK220245.1 | - | 11 | 1216 | |||

Galns
Genomic Map