siRNA Details
| Gene Symbol: Hnrnpdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016690.4 | NP_057899.2 | 3 | 794 | ||
| GenBank mRNA | AB017020.1 | - | 3 | 500 | ||
| AK011581.1 | - | 3 | 483 | |||
| AK011767.1 | - | 3 | 483 | |||
| AK172629.1 | - | 3 | 453 | |||
| BC021374.1 | - | 3 | 452 | 5039077 | ||
| CX219512.1 | - | 2 | 169 | |||

Hnrnpdl
Genomic Map