siRNA Details
| Gene Symbol: Mink1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045959.1 | NP_001039424.1 | 4 | 387 | ||
| NM_001045964.1 | NP_001039429.1 | 4 | 387 | |||
| NM_016713.2 | NP_057922.2 | 4 | 387 | |||
| NM_176893.2 | NP_795712.2 | 4 | 387 | |||
| GenBank mRNA | AB035697.1 | - | 4 | 372 | ||
| AB041925.1 | - | 4 | 373 | |||
| BC052474.1 | - | 4 | 251 | 6833893 | ||
| BC079549.1 | - | 4 | 245 | |||
| CB245214.1 | - | 4 | 251 | |||
| U49949.1 | - | 2 | 108 | |||

Mink1
Genomic Map