siRNA Details
| Gene Symbol: Ramp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168392.1 | NP_001161864.1 | 2 | 360 | ||
| NM_016894.3 | NP_058590.1 | 2 | 360 | |||
| NM_178401.3 | NP_848488.2 | 2 | 360 | |||
| GenBank mRNA | AB019046.1 | - | 2 | 171 | ||
| AF146522.1 | - | 2 | 152 | |||
| AF209904.1 | - | 2 | 131 | |||
| AJ250489.1 | - | 2 | 152 | |||
| AK144331.1 | - | 2 | 360 | |||
| AK157896.1 | - | 2 | 150 | |||
| AK165238.1 | - | 2 | 136 | |||
| BC012644.1 | - | 2 | 117 | 4207524 | ||
| BY707433.1 | - | 2 | 141 | |||
| BY733497.1 | - | 2 | 360 | |||

Ramp1
Genomic Map