siRNA Details
| Gene Symbol: Rif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175238.5 | NP_780447.4 | 7 | 736 | ||
| GenBank mRNA | AY428571.2 | - | 7 | 720 | ||
| AY585206.1 | - | 6 | 729 | |||
| JX255737.1 | - | 6 | 672 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rif1| Gene Symbol: Rif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175238.5 | NP_780447.4 | 7 | 736 | ||
| GenBank mRNA | AY428571.2 | - | 7 | 720 | ||
| AY585206.1 | - | 6 | 729 | |||
| JX255737.1 | - | 6 | 672 | |||