siRNA Details
| Gene Symbol: Tmem141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040130.3 | NP_001035219.2 | 4 | 243 | ||
| NM_001109993.1 | NP_001103463.1 | 3 | 505 | |||
| GenBank mRNA | AK004380.1 | - | 4 | 245 | ||
| BC048537.1 | - | 3 | 440 | 6532162 | ||
| BY704611.1 | - | 4 | 244 | |||
| CD774285.1 | - | 2 | 563 | |||

Tmem141
Genomic Map