siRNA Details
| Gene Symbol: R3hcc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177464.4 | NP_803415.1 | 6 | 2236 | ||
| GenBank mRNA | AK037718.1 | - | 2 | 1832 | ||
| AK077281.1 | - | 3 | 201 | |||
| AK082926.1 | - | 6 | 2237 | |||
| AK147893.1 | - | 6 | 2220 | |||
| BC038935.1 | - | 6 | 2212 | 5035077 | ||

R3hcc1l
Genomic Map