siRNA Details
| Gene Symbol: Ankrd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024204.6 | NP_077166.4 | 2 | 458 | ||
| GenBank mRNA | AK012869.1 | - | 2 | 471 | ||
| AK017360.1 | - | 2 | 459 | |||
| AK017408.1 | - | 2 | 343 | |||
| AK037242.1 | - | 2 | 341 | |||
| BC003460.1 | - | 2 | 320 | 2648395 | ||

Ankrd22
Genomic Map