siRNA Details
| Gene Symbol: Tmem38b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028053.2 | NP_082329.1 | 3 | 458 | ||
| GenBank mRNA | AB261159.1 | - | 3 | 394 | ||
| AK005479.1 | - | 3 | 441 | |||
| AK136131.1 | - | 2 | 208 | |||
| AK150377.1 | - | 3 | 436 | |||
| AK150892.1 | - | 3 | 436 | |||
| AK153075.1 | - | 3 | 423 | |||
| BC011072.1 | - | 3 | 436 | 3978742 | ||

Tmem38b
Genomic Map