siRNA Details
| Gene Symbol: Pvr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027514.2 | NP_081790.1 | 8 | 1293 | ||
| GenBank mRNA | AB195430.1 | - | 8 | 1250 | ||
| AJ563420.1 | - | 8 | 1181 | |||
| AK038747.1 | - | 8 | 1264 | |||
| AK078344.1 | - | 8 | 1260 | |||
| AK080753.1 | - | 8 | 1264 | |||
| AK165873.1 | - | 8 | 1283 | |||
| BC013673.1 | - | 8 | 1250 | 4188634 | ||
| BC032283.1 | - | 8 | 1253 | 5363187 | ||
| BC138219.1 | - | 8 | 1238 | 8861238 | ||
| BC138220.1 | - | 8 | 1238 | 8861239 | ||
| U35836.1 | - | 7 | 1212 | |||

Pvr
Genomic Map