siRNA Details
| Gene Symbol: D1Ertd622e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133825.3 | NP_598586.1 | 3 | 853 | ||
| GenBank mRNA | AK135656.1 | - | 3 | 799 | ||
| AK137265.1 | - | 3 | 1612 | |||
| AK152370.1 | - | 3 | 825 | |||
| AK162223.1 | - | 3 | 797 | |||
| AK168549.1 | - | 3 | 828 | |||
| BC019375.1 | - | 2 | 710 | 5008599 | ||
| BC023951.1 | - | 3 | 822 | 5320263 | ||
| BC034088.1 | - | 3 | 1316 | |||

D1Ertd622e
Genomic Map