siRNA Details
| Gene Symbol: Echdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254754.1 | NP_001241683.1 | 7, 8 | 787 | ||
| NM_026728.4 | NP_081004.2 | 8, 9 | 855 | |||
| GenBank mRNA | AK005030.1 | - | 8, 9 | 796 | ||
| AK011365.1 | - | 7, 8 | 733 | |||
| AK153878.1 | - | 8, 9 | 830 | |||
| AK160482.1 | - | 6, 7 | 558 | |||
| AK166965.1 | - | 8, 9 | 797 | |||
| BC025454.1 | - | 4, 5 | 339 | 5352625 | ||

Echdc2
Genomic Map