siRNA Details
| Gene Symbol: Echdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254754.1 | NP_001241683.1 | 9 | 889 | ||
| NM_026728.4 | NP_081004.2 | 10 | 957 | |||
| GenBank mRNA | AI839329.1 | - | 1 | 362 | ||
| AK005030.1 | - | 10 | 898 | |||
| AK011365.1 | - | 9 | 835 | |||
| AK153878.1 | - | 10 | 932 | |||
| AK160482.1 | - | 8 | 660 | |||
| AK166388.1 | - | 10 | 929 | |||
| AK166965.1 | - | 10 | 899 | |||
| BC025104.1 | - | 10 | 903 | 3591648 | ||
| BC025454.1 | - | 6 | 441 | 5352625 | ||

Echdc2
Genomic Map