siRNA Details
| Gene Symbol: Cenpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134046.5 | NP_598807.2 | 5 | 1022 | ||
| GenBank mRNA | AK013198.1 | - | 5 | 694 | ||
| AK018444.1 | - | 5 | 667 | |||
| AK029226.1 | - | 5 | 1023 | |||
| AK036089.1 | - | 4 | 527 | |||
| AK134266.1 | - | 5 | 694 | |||
| BC023148.1 | - | 5 | 665 | 4011571 | ||
| BC034399.1 | - | 5 | 681 | 4481047 | ||
| BC036278.1 | - | 5 | 665 | |||

Cenpo
Genomic Map