siRNA Details
| Gene Symbol: D16Ertd519e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139678.1 | - | 2 | 167 | ||
| BG068156.2 | - | 2 | 443 | |||
| CO811502.1 | - | 2 | 175 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D16Ertd519e| Gene Symbol: D16Ertd519e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139678.1 | - | 2 | 167 | ||
| BG068156.2 | - | 2 | 443 | |||
| CO811502.1 | - | 2 | 175 | |||