siRNA Details
| Gene Symbol: Zwint | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293683.1 | NP_001280612.1 | 5, 6 | 559 | ||
| NM_001293684.1 | NP_001280613.1 | 5, 6 | 559 | |||
| NM_025635.4 | NP_079911.1 | 5, 6 | 559 | |||
| GenBank mRNA | AK004305.1 | - | 5, 6 | 511 | ||
| AK008144.1 | - | 5, 6 | 509 | |||
| AK013556.1 | - | 5, 6 | 494 | |||
| AK075977.1 | - | 5, 6 | 505 | |||
| AK082215.1 | - | 5, 6 | 508 | |||
| AK134380.1 | - | 5, 6 | 511 | |||
| AK170637.1 | - | 1, 2 | 51 | |||
| BC013559.1 | - | 5, 6 | 477 | |||
| BC027100.1 | - | 5, 6 | 516 | 5359154 | ||
| BC034870.1 | - | 5, 6 | 488 | 1382192 | ||

Zwint
Genomic Map