siRNA Details
| Gene Symbol: D19Bwg1357e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050829.1 | - | 17 | 1831 | ||
| AK088081.1 | - | 17 | 1826 | |||
| AK141893.1 | - | 17 | 1829 | |||
| AK172872.1 | - | 13 | 1337 | |||
| BC041111.1 | - | 16 | 1809 | |||
| BC145533.1 | - | 17 | 1820 | |||

D19Bwg1357e
Genomic Map