siRNA Details
| Gene Symbol: Tomm40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109748.1 | NP_001103218.1 | 7 | 983 | ||
| NM_016871.2 | NP_058567.2 | 8 | 885 | |||
| GenBank mRNA | AF043249.1 | - | 7 | 661 | ||
| AF316403.1 | - | 7 | 818 | |||
| AF316404.1 | - | 7 | 818 | |||
| AK142575.1 | - | 7 | 579 | |||
| AK150019.1 | - | 7 | 980 | |||
| AK163759.1 | - | 8 | 869 | |||
| AK164570.1 | - | 8 | 869 | |||
| AK165173.1 | - | 8 | 880 | |||
| AK170991.1 | - | 8 | 869 | |||
| BC100452.1 | - | 8 | 873 | 6532792 | ||

Tomm40
Genomic Map