siRNA Details
| Gene Symbol: Reck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016678.2 | NP_057887.2 | 18 | 2413 | ||
| GenBank mRNA | AK042266.1 | - | 19 | 2592 | ||
| AK143657.1 | - | 18 | 2392 | |||
| AK157885.1 | - | 18 | 2402 | |||
| BC053025.1 | - | 18 | 2352 | |||
| BC132197.1 | - | 18 | 2441 | 40130474 | ||
| BC138065.1 | - | 18 | 2441 | 8861084 | ||
| BC145133.1 | - | 18 | 2442 | |||

Reck
Genomic Map