siRNA Details
| Gene Symbol: Insm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016889.3 | NP_058585.2 | 1 | 1022 | ||
| GenBank mRNA | BC051240.1 | - | 1 | 1098 | 6469481 | |
| BC082809.1 | - | 1 | 908 | 5720418 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Insm1| Gene Symbol: Insm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016889.3 | NP_058585.2 | 1 | 1022 | ||
| GenBank mRNA | BC051240.1 | - | 1 | 1098 | 6469481 | |
| BC082809.1 | - | 1 | 908 | 5720418 | ||