siRNA Details
| Gene Symbol: Pmm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016881.2 | NP_058577.1 | 6 | 546 | ||
| GenBank mRNA | AF043514.1 | - | 6 | 546 | ||
| AK002858.1 | - | 6 | 488 | |||
| AK003334.1 | - | 6 | 504 | |||
| AK005074.1 | - | 6 | 528 | |||
| AK134850.1 | - | 498 | ||||
| AK145402.1 | - | 6 | 498 | |||
| AK149116.1 | - | 6 | 508 | |||
| BC037101.1 | - | 6 | 523 | |||
| BC046325.1 | - | 6 | 494 | 6395635 | ||
| BC094448.1 | - | 6 | 512 | 4238664 | ||

Pmm2
Genomic Map