siRNA Details
Gene Symbol: Atxn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016843.3 | NP_058539.2 | 1 | 183 | ||
GenBank mRNA | AK002504.1 | - | 1 | 292 | ||
AK003530.1 | - | 1 | 110 | |||
AK013742.1 | - | 1 | 112 | |||
AK047170.1 | - | 1 | 233 | |||
AK049706.1 | - | 1 | 227 | |||
AK077738.1 | - | 1 | 185 | |||
AK078459.1 | - | 1 | 263 | |||
AK153271.1 | - | 1 | 185 | |||
AK161950.1 | - | 200 | ||||
AK165941.1 | - | 1 | 208 | |||
AK166899.1 | - | 1 | 268 | |||
AK170507.1 | - | 1 | 200 | |||
AK170751.1 | - | 1 | 209 | |||
BC016410.1 | - | 1 | 193 | 3968016 | ||
BC046802.1 | - | 1 | 319 | 6404607 |
