siRNA Details
| Gene Symbol: Rdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001150749.1 | NP_001144221.1 | 4 | 1047 | ||
| NM_017473.4 | NP_059501.1 | 4 | 1019 | |||
| GenBank mRNA | AB015165.1 | - | 4 | 1135 | ||
| AB032058.1 | - | 4 | 927 | |||
| AF056194.1 | - | 4 | 1024 | |||
| BC024603.1 | - | 4 | 997 | 4196075 | ||
| BC092255.1 | - | 4 | 973 | 5100891 | ||
| BC093514.1 | - | 4 | 961 | 5132290 | ||
| CB951082.1 | - | 1 | 146 | |||

Rdh7
Genomic Map