siRNA Details
| Gene Symbol: Rdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001150749.1 | NP_001144221.1 | 3 | 776 | ||
| NM_017473.4 | NP_059501.1 | 3 | 748 | |||
| GenBank mRNA | AB015165.1 | - | 3 | 864 | ||
| AB032058.1 | - | 3 | 656 | |||
| AF056194.1 | - | 3 | 753 | |||
| BC024603.1 | - | 3 | 726 | 4196075 | ||
| BC092255.1 | - | 3 | 702 | 5100891 | ||
| BC093514.1 | - | 3 | 690 | 5132290 | ||

Rdh7
Genomic Map